accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52611-08-2

IUPAC Name: palladium(2+) bis(methyldiphenylphosphane) dichloride
Synonyms: palladium(2+) bis(methyldiphenylphosphane) dichloride
Molecular Weight (g/mol): 577.76
Molecular Formula: C26H26Cl2P2Pd
InChi Key: GTSFBKGHQLDFHU-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].CP(C1=CC=CC=C1)C1=CC=CC=C1.CP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results