accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHOSSHONOHSSCu

CAS RN 52611-57-1

IUPAC Name: copper(2+) bis([bis(2-hydroxyethyl)carbamothioyl]sulfanide)
Synonyms: copper(2+) bis([bis(2-hydroxyethyl)carbamothioyl]sulfanide)
Molecular Weight (g/mol): 424.07
Molecular Formula: C10H20CuN2O4S4
InChi Key: CWZKOKSQKMDNMW-UHFFFAOYSA-L
SMILES: [Cu++].OCCN(CCO)C([S-])=S.OCCN(CCO)C([S-])=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results