accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5266-85-3

IUPAC Name: 2-methyl-6-(propan-2-yl)aniline
Synonyms: 2-isopropyl-6-methylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: DDTKYVBFPULMGN-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results