accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 527-20-8

IUPAC Name: pentachloroaniline
Synonyms: pentachloroaniline
Molecular Weight (g/mol): 265.34
Molecular Formula: C6H2Cl5N
InChi Key: KHCZSJXTDDHLGJ-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C(Cl)=C(Cl)C(Cl)=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results