accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOO

CAS RN 527-31-1

IUPAC Name: cyclohexane-1,2,3,4,5,6-hexone
Synonyms: cyclohexanehexone
Molecular Weight (g/mol): 168.06
Molecular Formula: C6O6
InChi Key: PKRGYJHUXHCUCN-UHFFFAOYSA-N
SMILES: O=C1C(=O)C(=O)C(=O)C(=O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)