accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CO

CAS RN 527-61-7

IUPAC Name: 2,6-dimethylcyclohexa-2,5-diene-1,4-dione
Synonyms: 2,6-dimethylcyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: SENUUPBBLQWHMF-UHFFFAOYSA-N
SMILES: CC1=CC(=O)C=C(C)C1=O
Percent Purity 
  • (3)
  • (1)
  • (4)
Quantity 
  • (4)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)