accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 52709-87-2

IUPAC Name: 4'-butoxy-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-butoxy-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 251.33
Molecular Formula: C17H17NO
InChi Key: KPQVQWUNELODQE-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results