accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5271-26-1

IUPAC Name: 2-phenylpiperazine
Synonyms: 2-phenylpiperazine
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: RIMRLBGNCLMSNH-UHFFFAOYNA-N
SMILES: C1CNC(CN1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results