accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52721-69-4

IUPAC Name: 2-(2-fluorophenyl)ethan-1-aminium
Synonyms: 2-(2-fluorophenyl)ethanaminium
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H11FN
InChi Key: RIKUOLJPJNVTEP-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=CC=C1F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)