accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHH3CH3CNHO

CAS RN 52722-86-8

IUPAC Name: 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol
Synonyms: 1-(2-hydroxyethyl)-2,2,6,6-tetramethylpiperidin-4-ol
Molecular Weight (g/mol): 201.31
Molecular Formula: C11H23NO2
InChi Key: STEYNUVPFMIUOY-UHFFFAOYSA-N
SMILES: CC1(C)CC(O)CC(C)(C)N1CCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results