accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52768-17-9

IUPAC Name: 4-(1H-pyrrol-1-yl)aniline
Synonyms: 4-(pyrrol-1-yl)aniline
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: NHLHWHRXMZZWGA-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results