accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFON

CAS RN 52771-22-9

IUPAC Name: 3-(trifluoromethoxy)benzonitrile
Synonyms: 3-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: DCZAPXGEZYVQNX-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(=C1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results