accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOCH3

CAS RN 5279-51-6

IUPAC Name: N-(4-methoxyphenyl)-N-methylformamide
Synonyms: N-(4-methoxyphenyl)-N-methylformamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: AOXIKOMIXYQQLL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N(C)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results