accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCH2H2COH

CAS RN 528-43-8

IUPAC Name: 5,5'-bis(prop-2-en-1-yl)-[1,1'-biphenyl]-2,2'-diol
Synonyms: magnolol
Molecular Weight (g/mol): 266.34
Molecular Formula: C18H18O2
InChi Key: VVOAZFWZEDHOOU-UHFFFAOYSA-N
SMILES: OC1=CC=C(CC=C)C=C1C1=CC(CC=C)=CC=C1O
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)