accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 528-75-6

IUPAC Name: 2,4-dinitrobenzaldehyde
Synonyms: 2,4-dinitrobenzaldehyde
Molecular Weight (g/mol): 196.12
Molecular Formula: C7H4N2O5
InChi Key: ZILXIZUBLXVYPI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=O)C(=C1)[N+]([O-])=O
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)