accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOO

CAS RN 52817-12-6

IUPAC Name: 6-bromo-4-oxo-4H-chromene-3-carbaldehyde
Synonyms: 6-bromo-4-oxochromene-3-carbaldehyde
Molecular Weight (g/mol): 253.05
Molecular Formula: C10H5BrO3
InChi Key: PCEZXSJBHMOQFT-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OC=C(C=O)C(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)