accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHN

CAS RN 52818-63-0

IUPAC Name: N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Synonyms: N-[(4-methoxyphenyl)methyl]pyridin-2-amine
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: SMJGJENXFSWIMQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(CNC2=CC=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results