accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52829-07-9

IUPAC Name: bis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
Synonyms: bis(3,3,5,5-tetramethylpiperidin-4-yl) decanedioate
Molecular Weight (g/mol): 480.73
Molecular Formula: C28H52N2O4
InChi Key: RWMYPXKVMUFMKS-UHFFFAOYSA-N
SMILES: CC1(C)CNCC(C)(C)C1OC(=O)CCCCCCCCC(=O)OC1C(C)(C)CNCC1(C)C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)