accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 528565-79-9

IUPAC Name: (2R,5R)-1-{2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl}-2,5-diphenylphospholane
Synonyms: (2R,5R)-1-{2-[(2R,5R)-2,5-diphenylphospholan-1-yl]ethyl}-2,5-diphenylphospholane
Molecular Weight (g/mol): 506.61
Molecular Formula: C34H36P2
InChi Key: VHHAZLMVLLIMHT-DXCZPEQUNA-N
SMILES: C(CP1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1)P1[C@H](CC[C@@H]1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)