accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3COS

CAS RN 52888-80-9

IUPAC Name: N,N-dipropyl(benzylsulfanyl)formamide
Synonyms: prosulfocarb
Molecular Weight (g/mol): 251.39
Molecular Formula: C14H21NOS
InChi Key: NQLVQOSNDJXLKG-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)SCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results