accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52898-49-4

IUPAC Name: N,4-dimethoxy-N-methylbenzamide
Synonyms: N,4-dimethoxy-N-methylbenzamide
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: FIRHLPVYIMMZPV-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results