accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 529-23-7

IUPAC Name: 2-aminobenzaldehyde
Synonyms: 2-aminobenzaldehyde
Molecular Weight (g/mol): 121.14
Molecular Formula: C7H7NO
InChi Key: FXWFZIRWWNPPOV-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)