accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 529-35-1

IUPAC Name: 5,6,7,8-tetrahydronaphthalen-1-ol
Synonyms: 5,6,7,8-tetrahydro-1-naphthol
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: SCWNNOCLLOHZIG-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1CCCC2
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)