accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOH

CAS RN 529-84-0

IUPAC Name: 6,7-dihydroxy-4-methyl-2H-chromen-2-one
Synonyms: 6,7-dihydroxy-4-methylcoumarin
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: KVOJTUXGYQVLAJ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC(O)=C(O)C=C12
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results