accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 5291-77-0

IUPAC Name: 1-benzylpyrrolidin-2-one
Synonyms: N-benzyl-2-pyrrolidinone
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: LVUQCTGSDJLWCE-UHFFFAOYSA-N
SMILES: O=C1CCCN1CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)