accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3NOO

CAS RN 5292-45-5

IUPAC Name: 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate
Synonyms: 1,4-dimethyl 2-nitrobenzene-1,4-dicarboxylate
Molecular Weight (g/mol): 239.18
Molecular Formula: C10H9NO6
InChi Key: PAYWCKGMOYQZAW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C(=O)OC)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results