accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 52927-22-7

IUPAC Name: 6-hydroxynaphthalene-2-carbonitrile
Synonyms: 6-hydroxynaphthalene-2-carbonitrile
Molecular Weight (g/mol): 169.18
Molecular Formula: C11H7NO
InChi Key: WKTNIBWKHNIPQR-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=C(C=C2C=C1)C#N
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results