accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COPOOHHOH3CCH3OPOOHOCH3H3C

CAS RN 52933-00-3

IUPAC Name: (propan-2-yloxy)phosphonic acid; bis(propan-2-yloxy)phosphinic acid
Synonyms: phosphorylisopropane diisopropylphosphate
Molecular Weight (g/mol): 322.23
Molecular Formula: C9H24O8P2
InChi Key: RTIXFJOJNSXSOB-UHFFFAOYSA-N
SMILES: CC(C)OP(O)(O)=O.CC(C)OP(O)(=O)OC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results