accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52938-94-0

IUPAC Name: 5-(4-cyanophenyl)furan-2-carboxylic acid
Synonyms: 5-(4-cyanophenyl)furan-2-carboxylic acid
Molecular Weight (g/mol): 213.19
Molecular Formula: C12H7NO3
InChi Key: QSWFKKSKRNCSDY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results