accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 52943-21-2

IUPAC Name: 2-chloro-N,N-dimethylpyridine-3-carboxamide
Synonyms: 2-chloro-N,N-dimethylpyridine-3-carboxamide
Molecular Weight (g/mol): 184.62
Molecular Formula: C8H9ClN2O
InChi Key: QNZRJGJNLOMEGJ-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=C(Cl)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results