accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFONH2

CAS RN 529512-81-0

IUPAC Name: 2-fluoro-6-methoxybenzamide
Synonyms: 2-fluoro-6-methoxybenzamide
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: BDJNFURQDZEPLK-UHFFFAOYSA-N
SMILES: COC1=C(C(N)=O)C(F)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results