CAS RN 52997-43-0
IUPAC Name:
7-(bromomethyl)pentadecane
Synonyms:
7-(bromomethyl)pentadecane
Molecular Weight (g/mol):
305.34
Molecular Formula:
C16H33Br
InChi Key:
RWEKWRQKAHQYNE-UHFFFAOYNA-N
SMILES:
CCCCCCCCC(CBr)CCCCCC
1
–
1
of
1
results
1
7-(Bromomethyl)pentadecane 95.0+%, TCI America™
CAS: 52997-43-0 Molecular Formula: C16H33Br Molecular Weight (g/mol): 305.344 MDL Number: MFCD00136231 InChI Key: RWEKWRQKAHQYNE-UHFFFAOYSA-N PubChem CID: 18459580 IUPAC Name: 7-(bromomethyl)pentadecane SMILES: CCCCCCCCC(CCCCCC)CBr