accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53-03-2

IUPAC Name: (1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-7,10-dione
Synonyms: prednisone metacortandracin hostacortin enkorton hoechst brand of prednisone 1,2-dehydrocortisone incocortyl cutason cortidelt pronisone
Molecular Weight (g/mol): 358.43
Molecular Formula: C21H26O5
InChi Key: XOFYZVNMUHMLCC-ZPOLXVRWSA-N
SMILES: C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Grade 
  • (1)
  • (6)
Percent Purity 
  • (6)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (6)
  • (1)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)