accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(R)OH(S)O

CAS RN 53-16-7

IUPAC Name: (3aS,3bR,9bS,11aS)-7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
Synonyms: estrone folipex (9 β)-isomer estrone tokokin estrone, (9 β)-isomer wehgen oestrone preparation folisan solliculin folliculine benzoate
Molecular Weight (g/mol): 270.37
Molecular Formula: C18H22O2
InChi Key: DNXHEGUUPJUMQT-CBZIJGRNSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CCC2=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)