CAS RN 53-21-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cocaine Hydrochloride Solution, 1.0 mg/mL in Methanol, Analytical Standard, For Drug Analysis, MilliporeSigma™ Supelco™
Cocaine is mostly commonly available as cocaine hydrochloride which is a white to off-white powder. It is taken by intranasal route. In some surgeries it is used as topical anaesthetic drug.