accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(R)(S)OH(R)H3C(S)O

CAS RN 53-43-0

IUPAC Name: (3aS,3bR,7S,9aR,9bS,11aS)-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
Synonyms: dehydroepiandrosterone DHEA prasterone
Molecular Weight (g/mol): 288.43
Molecular Formula: C19H28O2
InChi Key: FMGSKLZLMKYGDP-USOAJAOKSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CCC2=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (5)
  • (3)
Grade 
  • (5)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (5)
  • (3)
  • (3)
  • (5)
  • (5)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)