accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ON

CAS RN 530-40-5

IUPAC Name: N,N-diethylpyridine-4-carboxamide
Synonyms: N,N-diethylpyridine-4-carboxamide
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: CDQAJSRQKGFYDY-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=NC=C1
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results