accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClN

CAS RN 530-64-3

IUPAC Name: hydrogen quinoline chloride
Synonyms: hydrogen cinch chloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C9H8ClN
InChi Key: PSXRWZBTVAZNSF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C1=CC=C2N=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results