accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 530-75-6

IUPAC Name: 2-[2-(acetyloxy)benzoyloxy]benzoic acid
Synonyms: acetylsalicylsalicylic acid
Molecular Weight (g/mol): 300.27
Molecular Formula: C16H12O6
InChi Key: DDSFKIFGAPZBSR-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC=C1C(=O)OC1=CC=CC=C1C(O)=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results