accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3C

CAS RN 5306-85-4

IUPAC Name: 3,6-dimethoxy-hexahydrofuro[3,2-b]furan
Synonyms: dimethyl isosorbide
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: MEJYDZQQVZJMPP-UHFFFAOYNA-N
SMILES: COC1COC2C(COC12)OC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (5)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)