accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5307-03-9

IUPAC Name: 2-chloro-5-methylbenzene-1,4-diamine
Synonyms: 2-chloro-5-methylbenzene-1,4-diamine
Molecular Weight (g/mol): 156.61
Molecular Formula: C7H9ClN2
InChi Key: CPCPKQUNFFHAIZ-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(Cl)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results