accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOFFFF

CAS RN 53072-18-7

IUPAC Name: (2,3,4,5-tetrafluorophenyl)methanol
Synonyms: (2,3,4,5-tetrafluorophenyl)methanol
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: HLUZGUMMQYQHKJ-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results