accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH

CAS RN 5308-25-8

IUPAC Name: 1-ethylpiperazine
Synonyms: 1-ethylpiperazine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: WGCYRFWNGRMRJA-UHFFFAOYSA-N
SMILES: CCN1CCNCC1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)