accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(R)O(S)O(S)O(R)O

CAS RN 53081-25-7

IUPAC Name: (2R,3R,4S,5S,6R)-3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Synonyms: (2R,3R,4S,5S,6R)-3,4,5-tris(benzyloxy)-6-[(benzyloxy)methyl]oxan-2-ol
Molecular Weight (g/mol): 540.66
Molecular Formula: C34H36O6
InChi Key: OGOMAWHSXRDAKZ-BWNLSPMZSA-N
SMILES: O[C@@H]1O[C@H](COCC2=CC=CC=C2)[C@H](OCC2=CC=CC=C2)[C@H](OCC2=CC=CC=C2)[C@H]1OCC1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results