accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNN(Z)(E/Z)

CAS RN 531-52-2

IUPAC Name: (Z)-N'-(phenylamino)-N-(phenylimino)benzenecarboximidamide
Synonyms: (Z)-N'-(phenylamino)-N-(phenylimino)benzenecarboximidamide
Molecular Weight (g/mol): 300.37
Molecular Formula: C19H16N4
InChi Key: BEIHVSJTPTXQGB-QUHCWSQZSA-N
SMILES: N(\N=C(/N=NC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results