CAS RN 531-67-9
IUPAC Name:
1,1'-bipiperidine
Synonyms:
1,1'-bipiperidine
Molecular Weight (g/mol):
168.28
Molecular Formula:
C10H20N2
InChi Key:
QFDISQIDKZUABE-UHFFFAOYSA-N
SMILES:
C1CCN(CC1)N1CCCCC1
1
–
1
of
1
results
1
2,2'-Bipiperidine 98.0+%, TCI America™
CAS: 531-67-9 Molecular Formula: C10H20N2 Molecular Weight (g/mol): 168.284 MDL Number: MFCD01572939 InChI Key: CLBJZAWCBRAMRZ-UHFFFAOYSA-N PubChem CID: 415618 IUPAC Name: 2-piperidin-2-ylpiperidine SMILES: C1CCNC(C1)C2CCCCN2