accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SBr

CAS RN 53119-60-1

IUPAC Name: 2-bromo-4-methylthiophene
Synonyms: 2-bromo-4-methylthiophene
Molecular Weight (g/mol): 177.06
Molecular Formula: C5H5BrS
InChi Key: WQCPSNAAWLHQAE-UHFFFAOYSA-N
SMILES: CC1=CSC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results