accessibility menu, dialog, popup

UserName

Viewing profile as user:
OZrCe

CAS RN 53169-24-7

IUPAC Name: zirconium(4+) cerium(3+) oxidandiide
Synonyms: zirconium(4+) cerium(3+) oxidandiide
Molecular Weight (g/mol): 247.34
Molecular Formula: CeOZr
InChi Key: WPQBNJRIWONKBL-UHFFFAOYSA-N
SMILES: [O--].[Zr+4].[Ce+3]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)