accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53173-91-4

IUPAC Name: N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
Synonyms: N-[4-(4-bromophenyl)-1,3-thiazol-2-yl]acetamide
Molecular Weight (g/mol): 297.17
Molecular Formula: C11H9BrN2OS
InChi Key: CALREGGMKYUGIP-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC(=CS1)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results