accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSSNH2NN

CAS RN 5319-77-7

IUPAC Name: 5-(methylsulfanyl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(methylsulfanyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 147.21
Molecular Formula: C3H5N3S2
InChi Key: PCLAZAJARAIGGD-UHFFFAOYSA-N
SMILES: CSC1=NN=C(N)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results